(2-Hydroxyethyl)hydrazine ≥80.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: β-Hydroxyethylhydrazine

H1067-25G
TCH1067-25GEA 32 USD
TCH1067-25G
(2-Hydroxyethyl)hydrazine ≥80.0% (by titrimetric analysis)
(2-Hydroxyethyl)hydrazine

CAS Number: 109-84-2
MDL Number: MFCD00007623
Molecular Formula: C2H8N2O
Molecular Weight: 76.10
Purity/Analysis Method: 80.0% (T)
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2-Hydroxyethylhydrazine

Formula: C₂H₈N₂O
MW: 76.1 g/mol
Density: 1.12 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00007623
CAS Number: 109-84-2
UN: 2810
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/10/85/28971085.jpg

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR