Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid) ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9-Hydroxy-9-fluorenecarboxylic acid
H1004-25G
TCH1004-25GEA
97.44
USD
TCH1004-25G
Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid) ≥98.0% (by HPLC)
Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid)
CAS Number: 467-69-6
MDL Number: MFCD00001137
Molecular Formula: C14H10O3
Molecular Weight: 226.23
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: Flurenol
Formula:
C₁₄H₁₀O₃ MW: 226.23 g/mol |
MDL Number:
MFCD00001137 CAS Number: 467-69-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...