1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(4-Hydroxyphenyl)-5-mercaptotetrazole

H0896-25G
TCH0896-25GEA 102 USD
TCH0896-25G
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole ≥90.0% (by titrimetric analysis)
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole

CAS Number: 52431-78-4
MDL Number: MFCD00132898
Molecular Formula: C7H6N4OS
Molecular Weight: 194.21
Purity/Analysis Method: 90.0% (T)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: 1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol, 4-(5-Mercapto-1H-tetrazol-1-yl)phenol

Formula: C₇H₆N₄OS
Melting Pt: 162 °C
MDL Number: MFCD00132898
CAS Number: 52431-78-4
https://us.vwr.com/stibo/web/std.lang.all/09/18/28970918.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR