Heptanenitrile ≥98.0%

Supplier: TCI America

Synonyms: Hexyl Cyanide

H0883-25ML
TCH0883-25MLEA 101.5 USD
TCH0883-25ML
Heptanenitrile ≥98.0%
Heptanenitrile

CAS Number: 629-08-3
MDL Number: MFCD00039503
Molecular Formula: C7H13N
Molecular Weight: 111.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 58
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Hexyl Cyanide

Formula: C₇H₁₃N
MW: 111.19 g/mol
Boiling Pt: 183 °C
Density: 0.81 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00039503
CAS Number: 629-08-3
https://us.vwr.com/stibo/web/std.lang.all/09/05/28970905.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR