4-Heptylbenzoyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0792-25G
H0792-5G
TCH0792-5GEA
131.89
USD
TCH0792-5G
TCH0792-25G
4-Heptylbenzoyl chloride ≥98.0% (by titrimetric analysis)
4-Heptylbenzoyl chloride
CAS Number: 50606-96-7
MDL Number: MFCD00000702
Molecular Formula: C14H19ClO
Molecular Weight: 238.76
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 110
Specific Gravity (20/20): 1.03
Formula:
C₁₄H₁₉ClO MW: 238.76 g/mol Boiling Pt: 196 °C Density: 1.03 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00000702 CAS Number: 50606-96-7 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...