trans,trans-2,4-Hexadienal ≥80.0% (by GC)

Supplier: TCI America

H0581-25ML
TCH0581-25MLEA 85 USD
TCH0581-25ML
trans,trans-2,4-Hexadienal ≥80.0% (by GC)
trans,trans-2,4-Hexadienal

CAS Number: 142-83-6
MDL Number: MFCD00007004
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 67
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Sorbic Aldehyde

Formula: C₆H₈O
Density: 0.90 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00007004
CAS Number: 142-83-6
https://us.vwr.com/stibo/web/std.lang.all/05/94/28970594.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR