1,2-Diphenylhydrazine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Hydrazobenzene

H0180-100G H0180-25G
TCH0180-100GEA 575.51 USD
TCH0180-100G TCH0180-25G
1,2-Diphenylhydrazine ≥98.0% (by HPLC)
1,2-Diphenylhydrazine

CAS Number: 122-66-7
MDL Number: MFCD00003012
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,2-Diphenylhydrazine

Formula: C₁₂H₁₂N₂
Melting Pt: 127 °C
MDL Number: MFCD00003012
CAS Number: 122-66-7
https://us.vwr.com/stibo/web/std.lang.all/02/28/28970228.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR