Guanidine hydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Guanidine Hydrobromide
G0449-5G
G0449-1G
TCG0449-1GEA
57.84
USD
TCG0449-1G
TCG0449-5G
Guanidine hydrobromide ≥98.0% (by titrimetric analysis)
Guanidine hydrobromide
CAS Number: 19244-98-5
MDL Number: MFCD03094062
Molecular Formula: CH5N3
Molecular Weight: 139.98
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 191
Formula:
CH₅N₃ MW: 139.98 g/mol Melting Pt: 191 °C |
MDL Number:
MFCD03094062 CAS Number: 19244-98-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...