Gaboxadol hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,5,6,7-Tetrahydroisoxazolo[5,4-c]pyridin-3-ol Hydrochloride

G0405-25MG
TCG0405-25MGEA 161.13 USD
TCG0405-25MG
Gaboxadol hydrochloride ≥98.0% (by HPLC)
Gaboxadol hydrochloride

CAS Number: 85118-33-8
MDL Number: MFCD00055180
Molecular Formula: C6H8N2O2
Molecular Weight: 176.60
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 236


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 4,5,6,7-Tetrahydroisoxazolo[5,4-c]pyridin-3-ol Hydrochloride

Formula: C₆H₈N₂O₂
MW: 176.60 g/mol
Melting Pt: 236 °C
MDL Number: MFCD00055180
CAS Number: 85118-33-8
https://us.vwr.com/stibo/web/std.lang.all/99/98/28969998.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR