Gold's reagent ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
[[[(Dimethylamino)methylene]amino]methylene]dimethylammonium Chloride
G0393-5G
G0393-25G
TCG0393-25GEA
191.84
USD
TCG0393-25G
TCG0393-5G
Gold's reagent ≥97.0% (by HPLC, titration analysis)
Gold's reagent
CAS Number: 20353-93-9
MDL Number: MFCD00011793
Molecular Formula: C6H14ClN3
Molecular Weight: 163.65
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 99
Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: [[[(Dimethylamino)methylene]amino]methylene]dimethylammonium Chloride
Formula:
C₆H₁₄ClN₃ MW: 163.65 g/mol Melting Pt: 99 °C |
MDL Number:
MFCD00011793 CAS Number: 20353-93-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...