Glycerol formal (=1,3-dioxan-5-ol) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-Hydroxy-1,3-dioxane
, 1,3-Dioxan-5-ol
G0391-500MG
TCG0391-500MGEA
79.17
USD
TCG0391-500MG
Glycerol formal (=1,3-dioxan-5-ol) ≥98.0% (by GC)
Glycerol formal (=1,3-dioxan-5-ol)
CAS Number: 4740-78-7
MDL Number: MFCD10039829
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 93
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 5-Hydroxy-1,3-dioxane, 1,3-Dioxan-5-ol
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 194 °C Density: 1.23 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD10039829 CAS Number: 4740-78-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...