GlcNAc-β-(1-3)GalNAc-ɑ-pNP ≥98.0% (by HPLC)

Supplier: TCI America

G0376-5MG
TCG0376-5MGEA 387 USD
TCG0376-5MG
GlcNAc-β-(1-3)GalNAc-ɑ-pNP ≥98.0% (by HPLC)
GlcNAc-beta-(1-3)GalNAc-alpha-pNP

CAS Number: 125455-64-3
Molecular Formula: C22H31N3O13
Molecular Weight: 545.50
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C

Formula: C₂₂H₃₁N₃O₁₃
MW: 545.50 g/mol
CAS Number: 125455-64-3
https://us.vwr.com/stibo/web/std.lang.all/99/79/28969979.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR