Gnetol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2,3',5',6-Tetrahydroxy-trans-stilbene

G0371-100MG
TCG0371-100MGEA 236 USD
TCG0371-100MG
Gnetol ≥97.0% (by GC)
Gnetol

CAS Number: 86361-55-9
MDL Number: MFCD12405548
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 271
Lambda max.: 337 nm (EtOH)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2,3',5',6-Tetrahydroxy-trans-stilbene

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
Melting Pt: 271 °C
MDL Number: MFCD12405548
CAS Number: 86361-55-9
https://us.vwr.com/stibo/web/std.lang.all/99/76/28969976.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR