GalNAcβ(1-4)GlcNAc-β-pNP ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-galactopyranosyl)-beta-D-glucopyranoside
CAS Number: 872578-72-8
Molecular Formula: C22H31N3O13
Molecular Weight: 545.50
Purity/Analysis Method:
Form: Crystal
Storage Temperature:
Product Categories: Synthetic Organic Chemistry, p-Nitrophenyl Glycosides, Bioscience, Disaccharides, Glycosides, p-Nitrophenyl Glycosides
Other Product Names / Synonyms: 4-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-galactopyranosyl)-beta-D-glucopyranoside
Formula:
C₂₂H₃₁N₃O₁₃ MW: 545.50 g/mol |
CAS Number:
872578-72-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...