GalNAcβ(1-3)Galɑ(1-4)Galβ(1-4)Glc-β-pNP ≥88.0% (by HPLC)

G0354-5MG
TCG0354-5MGEA 782.8 USD
TCG0354-5MG
GalNAcβ(1-3)Galɑ(1-4)Galβ(1-4)Glc-β-pNP ≥88.0% (by HPLC)
GalNAc beta(1-3)Gal alpha(1-4)Gal beta(1-4)Glc-beta-pNP

CAS Number: 1134635-03-2
Molecular Formula: C32H48N2O23
Molecular Weight: 828.73
Purity/Analysis Method: 88.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C

Formula: C₃₂H₄₈N₂O₂₃
MW: 828.73 g/mol
CAS Number: 1134635-03-2
https://us.vwr.com/stibo/web/std.lang.all/99/66/28969966.jpg

Order Now

Specification Test Results

Purity/Analysis Method 88.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR