GlcNAc-β-(1-3)GalNAc-ɑ-Thr ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: O-[2-Acetamido-2-deoxy-3-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-alpha-D-galactopyranosyl]-L-threonine

G0341-2MG
TCG0341-2MGEA 733 USD
TCG0341-2MG
GlcNAc-β-(1-3)GalNAc-ɑ-Thr ≥97.0% (by HPLC)
GlcNAc-beta-(1-3)GalNAc-alpha-Thr

CAS Number: 286959-52-2
Molecular Formula: C20H35N3O13
Molecular Weight: 525.51
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 60 deg (C=1, H2O)
Storage Temperature: 0°C


Product Categories: Bioscience, Glycoamino Acids, Disaccharides
Other Product Names / Synonyms: O-[2-Acetamido-2-deoxy-3-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-alpha-D-galactopyranosyl]-L-threonine

Formula: C₂₀H₃₅N₃O₁₃
MW: 525.51 g/mol
CAS Number: 286959-52-2

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR