Ganciclovir ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]guanine
CAS Number: 82410-32-0
MDL Number: MFCD00870588
Molecular Formula: C9H13N5O4
Molecular Weight: 255.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 250
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]guanine
Formula:
C₉H₁₃N₅O₄ .xH₂O MW: 255.23 g/mol Melting Pt: 250 °C |
MDL Number:
MFCD00870588 CAS Number: 82410-32-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...