Galantamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
G0293-100MG
G0293-1G
TCG0293-1GEA
530.85
USD
TCG0293-1G
TCG0293-100MG
Galantamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Galantamine hydrobromide
CAS Number: 1953-04-4
Molecular Formula: C17H21NO3
Molecular Weight: 368.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 256
Specific rotation [a]20/D: -95 deg (C=1.4, H2O)
Storage Temperature:
Formula:
C₁₇H₂₁NO₃ MW: 368.27 g/mol Melting Pt: 256 °C |
MDL Number:
MFCD00067672 CAS Number: 1953-04-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...