Glutaronitrile ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Pentanedinitrile

G0072-250G G0072-25G
TCG0072-25GEA 45.68 USD
TCG0072-25G TCG0072-250G
Glutaronitrile ≥96.0% (by GC)
Glutaronitrile

CAS Number: 544-13-8
MDL Number: MFCD00001970
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 286
Flash Point (°C): 110
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,3-Dicyanopropane, Pentanedinitrile, Trimethylene Dicyanide

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 286 °C
Density: 0.99 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001970
CAS Number: 544-13-8
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/97/59/28969759.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR