4-Fluoro-(3-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Synonyms: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid4-Fluoro-3-methoxycarbonylphenylboronic acid

F0951-1G F0951-5G
TCF0951-1GEA 84.36 USD
TCF0951-1G TCF0951-5G
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid

CAS Number: 874219-35-9
MDL Number: MFCD06801688
Molecular Formula: C8H8BFO4
Molecular Weight: 197.96
Form: Crystal
Color: White
Melting point (°C): 255


Other Product Names / Synonyms: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic Acid

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 255 °C
MDL Number: MFCD06801688
CAS Number: 874219-35-9
https://us.vwr.com/stibo/web/std.lang.all/94/81/28969481.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR