3-Fluoro-2-nitrophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
F0947-1G
F0947-5G
TCF0947-5GEA
111
USD
TCF0947-5G
TCF0947-1G
3-Fluoro-2-nitrophenol ≥98.0% (by GC, titration analysis)
3-Fluoro-2-nitrophenol
CAS Number: 385-01-3
MDL Number: MFCD07368753
Molecular Formula: C6H4FNO3
Molecular Weight: 157.10
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 38
Storage Temperature: 0-10°C
Formula:
C₆H₄FNO₃ MW: 157.10 g/mol Melting Pt: 38 °C |
MDL Number:
MFCD07368753 CAS Number: 385-01-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...