Fluorescein isothiocyanate dextran, M.W. ∼ 10000
Supplier: TCI America
Synonyms:
FITC Dextran
F0918-100MG
TCF0918-100MGEA
156.32
USD
TCF0918-100MG
Fluorescein isothiocyanate dextran, M.W. ∼ 10000
Fluorescein isothiocyanate dextran
CAS Number: 60842-46-8
MDL Number: MFCD00131092
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: 0-10°C
Other Product Names / Synonyms: FITC Dextran
MDL Number:
MFCD00131092 CAS Number: 60842-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...