Fludarabine monophosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Fluoroadenine 9-beta-D-Arabinofuranoside 5'-Monophosphate9-(beta-D-Arabinofuranosyl)-2-fluoroadenine 5'-Monophosphate2-F-ara-AMP

F0913-25MG F0913-100MG
TCF0913-100MGEA 85.23 USD
TCF0913-100MG TCF0913-25MG
Fludarabine monophosphate ≥98.0% (by HPLC, titration analysis)
Fludarabine monophosphate

CAS Number: 75607-67-9
MDL Number: MFCD00866418
Molecular Formula: C10H13FN5O7P
Molecular Weight: 365.21
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 203
Specific rotation [a]20/D: 12 deg (C=0.5, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleotides and Their Analogs
Other Product Names / Synonyms: 2-Fluoroadenine 9-beta-D-Arabinofuranoside 5'-Monophosphate, 9-(beta-D-Arabinofuranosyl)-2-fluoroadenine 5'-Monophosphate, 2-F-ara-AMP

Formula: C₁₀H₁₃FN₅O₇P
MW: 365.21 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00866418
CAS Number: 75607-67-9
https://us.vwr.com/stibo/web/std.lang.all/94/48/28969448.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR