Thiazole-5-carboxyaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 5-Thiazolecarboxaldehyde5-Formylthiazole

F0908-5G F0908-1G
TCF0908-5GEA 237.51 USD
TCF0908-5G TCF0908-1G
Thiazole-5-carboxyaldehyde ≥98.0% (by GC)
Thiazole-5-carboxyaldehyde

CAS Number: 1003-32-3
MDL Number: MFCD02179516
Molecular Formula: C4H3NOS
Molecular Weight: 113.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 98
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: 5-Thiazolecarboxaldehyde

Formula: C₄H₃NOS
MW: 113.14 g/mol
Boiling Pt: 94 °C
Density: 1.34 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD02179516
CAS Number: 1003-32-3
https://us.vwr.com/stibo/web/std.lang.all/94/42/28969442.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR