6-Fluoro-3-formylchromone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Fluorochromone-3-carboxaldehyde6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

F0905-1G
TCF0905-1GEA 25 USD
TCF0905-1G
6-Fluoro-3-formylchromone ≥98.0% (by GC, titration analysis)
6-Fluoro-3-formylchromone

CAS Number: 69155-76-6
MDL Number: MFCD00139060
Molecular Formula: C10H5FO3
Molecular Weight: 192.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 157


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoro-3-formylchromone, 6-Fluoro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Formula: C₁₀H₅FO₃
MW: 192.15 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00139060
CAS Number: 69155-76-6
https://us.vwr.com/stibo/web/std.lang.all/94/41/28969441.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR