Fluvoxamine maleate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone O-(2-Aminoethyl)oxime Maleate

F0858-1G F0858-5G
TCF0858-1GEA 159.24 USD
TCF0858-1G TCF0858-5G
Fluvoxamine maleate ≥98.0% (by HPLC, titration analysis)
Fluvoxamine maleate

CAS Number: 61718-82-9
MDL Number: MFCD00269809
Molecular Formula: C15H21F3N2O2
Molecular Weight: 434.41
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 122
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone O-(2-Aminoethyl)oxime Maleate

Formula: C₁₅H₂₁F₃N₂O₂
MW: 434.41 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00269809
CAS Number: 61718-82-9
https://us.vwr.com/stibo/web/std.lang.all/93/94/28969394.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR