3-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Fluoro-2-hydroxybenzoic acid

F0845-1G F0845-5G
TCF0845-1GEA 52.78 USD
TCF0845-1G TCF0845-5G
3-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)
3-Fluorosalicylic acid

CAS Number: 341-27-5
MDL Number: MFCD00153167
Molecular Formula: C7H5FO3
Molecular Weight: 156.11
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 148


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-2-hydroxybenzoic Acid

Formula: C₇H₅FO₃
MW: 156.11 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00153167
CAS Number: 341-27-5
https://us.vwr.com/stibo/web/std.lang.all/93/83/28969383.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR