Famciclovir ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate

F0842-5G F0842-500MG
TCF0842-5GEA 441.13 USD
TCF0842-5G TCF0842-500MG
Famciclovir ≥98.0% (by HPLC, titration analysis)
Famciclovir

CAS Number: 104227-87-4
MDL Number: MFCD00866964
Molecular Formula: C14H19N5O4
Molecular Weight: 321.34
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 105
Lambda max.: 310 nm (EtOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Prodrug Ingredients
Other Product Names / Synonyms: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate

Formula: C₁₄H₁₉N₅O₄
MW: 321.34 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00866964
CAS Number: 104227-87-4
https://us.vwr.com/stibo/web/std.lang.all/93/80/28969380.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR