Fasudil hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-(5-Isoquinolinesulfonyl)homopiperazine Hydrochloride5-(1-Homopiperazinyl)sulfonylisoquinoline Hydrochloride

F0839-100MG
TCF0839-100MGEA 100.45 USD
TCF0839-100MG
Fasudil hydrochloride ≥98.0% (by HPLC)
Fasudil hydrochloride

CAS Number: 105628-07-7
MDL Number: MFCD00943198
Molecular Formula: C14H17N3O2S
Molecular Weight: 327.83
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 222


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1-(5-Isoquinolinesulfonyl)homopiperazine Hydrochloride, 5-(1-Homopiperazinyl)sulfonylisoquinoline Hydrochloride

Formula: C₁₄H₁₇N₃O₂S
MW: 327.83 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00943198
CAS Number: 105628-07-7
https://us.vwr.com/stibo/web/std.lang.all/93/79/28969379.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR