1-(4-Fluorophenyl)-2-thiourea ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4-Fluorophenylthiourea N-(4-Fluorophenyl)thiourea

F0836-5G F0836-25G
TCF0836-5GEA 49 USD
TCF0836-5G TCF0836-25G
1-(4-Fluorophenyl)-2-thiourea ≥97.0% (by HPLC, total nitrogen)
1-(4-Fluorophenyl)-2-thiourea

CAS Number: 459-05-2
MDL Number: MFCD00041180
Molecular Formula: C7H7FN2S
Molecular Weight: 170.21
Purity/Analysis Method: 97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 160

Formula: (C₆H₄F)NHC(S)NH₂
MW: 170.21 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00041180
CAS Number: 459-05-2
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/93/76/28969376.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR