2-Fluorophenylsulfur pentafluoride ≥98.0% (by GC)

Supplier: TCI America

F0823-1G
TCF0823-1GEA 173.57 USD
TCF0823-1G
2-Fluorophenylsulfur pentafluoride ≥98.0% (by GC)
2-Fluorophenylsulphur pentafluoride

CAS Number: 864230-02-4
MDL Number: MFCD18909664
Molecular Formula: C6H4F6S
Molecular Weight: 222.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Storage Temperature: 0-10°C

Formula: C₆H₄F₆S
Boiling Pt: 94 °C
MDL Number: MFCD18909664
CAS Number: 864230-02-4
https://us.vwr.com/stibo/web/std.lang.all/93/62/28969362.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR