Fluvastatin sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid, sodium salt

F0820-5G F0820-1G
TCF0820-1GEA 105.97 USD
TCF0820-1G TCF0820-5G
Fluvastatin sodium salt hydrate ≥98.0% (by HPLC, total nitrogen)
Fluvastatin sodium salt hydrate

CAS Number: 93957-55-2
MDL Number: MFCD00929076
Molecular Formula: C24H26FNO4
Molecular Weight: 433.46
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C24H25FNNaO4 .xH2O
MW: 433.46 g/mol
MDL Number: MFCD00929076
CAS Number: 93957-55-2
https://us.vwr.com/stibo/web/std.lang.all/93/59/28969359.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR