(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal

F0819-25G F0819-5G
TCF0819-5GEA 164 USD
TCF0819-5G TCF0819-25G
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein ≥98.0% (by GC)
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein

CAS Number: 93957-50-7
MDL Number: MFCD03840863
Molecular Formula: C20H18FNO
Molecular Weight: 307.37
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 131


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal

Formula: C₂₀H₁₈FNO
MW: 307.37 g/mol
Melting Pt: 131 °C
MDL Number: MFCD03840863
CAS Number: 93957-50-7
https://us.vwr.com/stibo/web/std.lang.all/93/58/28969358.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR