3-Fluoro-N-methylbenzylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(3-Fluorophenyl)-N-methylmethanamine(3-Fluorobenzyl)methylamine

F0805-1G
TCF0805-1GEA 37 USD
TCF0805-1G
3-Fluoro-N-methylbenzylamine ≥97.0% (by GC, titration analysis)
3-Fluoro-N-methylbenzylamine

CAS Number: 90389-84-7
MDL Number: MFCD04623547
Molecular Formula: C8H10FN
Molecular Weight: 139.17
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 66
Specific Gravity (20/20): 1.05

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 184 °C
Density: 1,05 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD04623547
CAS Number: 90389-84-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR