2-Fluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Fluorobenzene-1-sulphonyl chloride

F0795-5G F0795-25G
TCF0795-5GEA 109.62 USD
TCF0795-5G TCF0795-25G
2-Fluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2-Fluorobenzenesulphonyl chloride

CAS Number: 2905-21-7
Molecular Formula: C6H4ClFO2S
Molecular Weight: 194.60
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 127
Flash Point (°C): 110
Freezing point (°C): 24

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 127 °C
Flash Pt: 110 °C
MDL Number: MFCD00042285
CAS Number: 2905-21-7
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/93/36/28969336.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR