3-Fluoro-4-methylbenzaldehyde ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 3-Fluoro-4-tolualdehyde 3-Fluoro-p-tolualdehyde 3-Fluoro-p-tolylaldehyde

F0792-5G
TCF0792-5GEA 92 USD
TCF0792-5G
3-Fluoro-4-methylbenzaldehyde ≥95.0% (by GC)
3-Fluoro-4-methylbenzaldehyde

CAS Number: 177756-62-6
MDL Number: MFCD01631532
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 82
Specific Gravity (20/20): 1.14

Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 206 °C
Density: 1.14 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD01631532
CAS Number: 177756-62-6
https://us.vwr.com/stibo/web/std.lang.all/93/33/28969333.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR