3-Fluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Fluorobenzene-1-sulphonyl chloride

F0790-5G F0790-25G
TCF0790-5GEA 43.54 USD
TCF0790-5G TCF0790-25G
3-Fluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-Fluorobenzenesulphonyl chloride

CAS Number: 701-27-9
MDL Number: MFCD00042286
Molecular Formula: C6H4ClFO2S
Molecular Weight: 194.60
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 113
Melting point (°C): 7
Flash Point (°C): 110
Specific Gravity (20/20): 1.47

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 113 °C
Melting Pt: 7 °C
Density: 1.47 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00042286
CAS Number: 701-27-9
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/93/31/28969331.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR