2-Formylselenophene ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 2-Selenophenecarboxaldehyde

F0782-1G
TCF0782-1GEA 168.41 USD
TCF0782-1G
2-Formylselenophene ≥98.0% (by GC) stabilized
2-Formylselenophene

(stabilized with HQ)
CAS Number: 25109-26-6
Molecular Formula: C5H4OSe
Molecular Weight: 159.05
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Specific Gravity (20/20): 1.67
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-Selenophenecarboxaldehyde

Formula: C₅H₄OSe
MW: 159.05 g/mol
Boiling Pt: 85 °C
Density: 1.67 (20 °C)
CAS Number: 25109-26-6
https://us.vwr.com/stibo/web/std.lang.all/93/23/28969323.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR