1-(2-Fluorophenyl)piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0728-25G
TCF0728-25GEA 79.17 USD
TCF0728-25G
1-(2-Fluorophenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(2-Fluorophenyl)piperazine

CAS Number: 1011-15-0
MDL Number: MFCD00040726
Molecular Formula: C10H13FN2
Molecular Weight: 180.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 93
Melting point (°C): 49
Flash Point (°C): 113

Formula: C₁₀H₁₃FN₂
MW: 180.23 g/mol
Boiling Pt: 93 °C
Melting Pt: 49 °C
Flash Pt: 113 °C
MDL Number: MFCD00040726
CAS Number: 1011-15-0
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR