4-Fluorphenylmagnesium bromide ∼19% ∼1.0 mol/L in THF
Supplier: TCI America
Synonyms:
4-Fluorophenylmagnesium bromide
F0673-250G
TCF0673-250GEA
104.55
USD
TCF0673-250G
4-Fluorphenylmagnesium bromide ∼19% ∼1.0 mol/L in THF
4-Fluorphenylmagnesium bromide
CAS Number: 352-13-6
MDL Number: MFCD00009667
Molecular Formula: C6H4BrFMg
Molecular Weight: 199.31
Flash Point (°C): -20
Formula:
C₆H₄BrFMg Flash Pt: –20 °C |
MDL Number:
MFCD00009667 CAS Number: 352-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...