Fluoroacetonitrile ≥98.0% (by GC)

Supplier: TCI America

F0665-5G
TCF0665-5GEA 164.87 USD
TCF0665-5G
Fluoroacetonitrile ≥98.0% (by GC)
Fluoroacetonitrile

CAS Number: 503-20-8
MDL Number: MFCD00009969
Molecular Formula: C2H2FN
Molecular Weight: 59.04
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Melting point (°C): -13
Flash Point (°C): -14
Specific Gravity (20/20): 1.08

Formula: C₂H₂FN
Boiling Pt: 82 °C
Melting Pt: –13 °C
Density: 1.08 (20 °C)
Flash Pt: –14 °C
MDL Number: MFCD00009969
CAS Number: 503-20-8
https://us.vwr.com/stibo/web/std.lang.all/92/12/28969212.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR