Fluorocyclohexane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclohexyl fluoride

F0587-5G
TCF0587-5GEA 222.19 USD
TCF0587-5G
Fluorocyclohexane ≥98.0% (by GC)
Fluorocyclohexane

CAS Number: 372-46-3
MDL Number: MFCD00021285
Molecular Formula: C6H11F
Molecular Weight: 102.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Melting point (°C): 13
Flash Point (°C): 5
Specific Gravity (20/20): 0.93

Formula: C₆H₁₁F
MW: 102.15 g/mol
Boiling Pt: 104 °C
Melting Pt: 13 °C
Density: 0.93 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00021285
CAS Number: 372-46-3
https://us.vwr.com/stibo/web/std.lang.all/91/45/28969145.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR