2-Fluoro-7,7,8,8-tetracyanoquinodimethane ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: FTCNQ

F0509-100MG
TCF0509-100MGEA 371 USD
TCF0509-100MG
2-Fluoro-7,7,8,8-tetracyanoquinodimethane ≥98.0% (by total nitrogen basis)
2-Fluoro-7,7,8,8-tetracyanoquinodimethane

CAS Number: 69857-37-0
Molecular Formula: C12H3FN4
Molecular Weight: 222.18
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 218


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: FTCNQ

Formula: C₁₂H₃FN₄
MW: 222.18 g/mol
Melting Pt: 218 °C
CAS Number: 69857-37-0
https://us.vwr.com/stibo/web/std.lang.all/90/75/28969075.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR