2,2-Difluoro-2-(fluorosulfonyl)acetic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

F0500-5G
TCF0500-5GEA 49.72 USD
TCF0500-5G
2,2-Difluoro-2-(fluorosulfonyl)acetic acid ≥95.0% (by titrimetric analysis)
2,2-Difluoro-2-(fluorosulphonyl)acetic acid

CAS Number: 1717-59-5
MDL Number: MFCD01320807
Molecular Formula: C2HF3O4S
Molecular Weight: 178.08
Purity/Analysis Method: 95.0% (T)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 110
Specific Gravity (20/20): 1.72

Formula: C₂HF₃O₄S
Boiling Pt: 153 °C
Density: 1.72 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01320807
CAS Number: 1717-59-5
https://us.vwr.com/stibo/web/std.lang.all/90/69/28969069.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR