α,α,α,2-Tetrafluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Fluorobenzotrifluoride 1-Fluoro-2-(trifluoromethyl)benzene

F0389-25G F0389-5G
TCF0389-5GEA 27.41 USD
TCF0389-5G TCF0389-25G
α,α,α,2-Tetrafluorotoluene ≥98.0% (by GC)
α,α,α,2-Tetrafluorotoluene

CAS Number: 392-85-8
MDL Number: MFCD00000374
Molecular Formula: C7H4F4
Molecular Weight: 164.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 17
Specific Gravity (20/20): 1.31


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-2-(trifluoromethyl)benzene, alpha,alpha,alpha,2-Tetrafluorotoluene

Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 116 °C
Density: 1.31 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00000374
CAS Number: 392-85-8
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/89/85/28968985.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR