3-Fluoro-o-xylene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1,2-Dimethyl-3-fluorobenzene
, 1-Fluoro-2,3-dimethylbenzene
F0332-5ML
TCF0332-5MLEA
59
USD
TCF0332-5ML
3-Fluoro-o-xylene ≥98.0% (by GC)
3-Fluoro-o-xylene
CAS Number: 443-82-3
MDL Number: MFCD00000323
Molecular Formula: C8H9F
Molecular Weight: 124.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 36
Specific Gravity (20/20): 1.01
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3-Dimethylfluorobenzene, 1-Fluoro-2,3-dimethylbenzene
Formula:
C₈H₉F MW: 124.16 g/mol Boiling Pt: 148 °C Density: 1.01 (20 °C) Flash Pt: 36 °C |
MDL Number:
MFCD00000323 CAS Number: 443-82-3 UN: 1993 ADR: 3,III |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...