2-Fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

F0217-25G
TCF0217-25GEA 62.93 USD
TCF0217-25G
2-Fluoropyridine ≥98.0% (by GC)
2-Fluoropyridine

CAS Number: 372-48-5
MDL Number: MFCD00006224
Molecular Formula: C5H4FN
Molecular Weight: 97.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 24
Specific Gravity (20/20): 1.13

Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 126 °C
Density: 1.13 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00006224
CAS Number: 372-48-5
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/88/23/28968823.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR