1-Fluoro-3-nitrobenzene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
3-Fluoronitrobenzene
F0110-25G
TCF0110-25GEA
109.62
USD
TCF0110-25G
1-Fluoro-3-nitrobenzene ≥97.0% (by GC)
1-Fluoro-3-nitrobenzene
CAS Number: 402-67-5
MDL Number: MFCD00007196
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 87
Specific Gravity (20/20): 1.33
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Boiling Pt: 198 °C Density: 1.33 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00007196 CAS Number: 402-67-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...