Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0%

Supplier: TCI America

Synonyms: 3-(Benzylamino)-3-oxopropionic Acid Ethyl EsterEthyl N-BenzylmalonamateN-Benzylmalonamic Acid Ethyl EsterEthyl (N-Benzylcarbamoyl)acetateEthyl 3-(benzylamino)-3-oxopropanoate(N-Benzylcarbamoyl)acetic Acid Ethyl Ester

E1098-1G E1098-5G
TCE1098-5GEA 387 USD
TCE1098-5G TCE1098-1G
Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0%
Ethyl-3-(benzylamino)-3-oxopropionate

CAS Number: 29689-63-2
Molecular Formula: C12H15NO3
Molecular Weight: 221.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 51


Other Product Names / Synonyms: 3-(Benzylamino)-3-oxopropionic Acid Ethyl Ester, Ethyl (N-Benzylcarbamoyl)acetate, (N-Benzylcarbamoyl)acetic Acid Ethyl Ester, Ethyl N-Benzylmalonamate, N-Benzylmalonamic Acid Ethyl Ester

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
Melting Pt: 51 °C
MDL Number: MFCD28128094
CAS Number: 29689-63-2
https://us.vwr.com/stibo/web/std.lang.all/84/39/28968439.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR