Ethyl-2-benzoylpropionate ≥98.0%
Supplier: TCI America
Synonyms:
2-Benzoylpropionic Acid Ethyl Ester
E1071-1G
E1071-5G
TCE1071-5GEA
486
USD
TCE1071-5G
TCE1071-1G
Ethyl-2-benzoylpropionate ≥98.0%
Ethyl-2-benzoylpropionate
CAS Number: 10488-87-6
MDL Number: MFCD00026882
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.09
Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 2-Benzoylpropionic Acid Ethyl Ester
Formula:
C₁₂H₁₄O₃ MW: 206.24 g/mol Boiling Pt: 96 °C Density: 1.09 (20 °C) |
MDL Number:
MFCD00026882 CAS Number: 10488-87-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...